siRNA Details
Gene Symbol: Nlrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ117583.1 | - | 2198 | |||
DQ117584.1 | - | 2 | 2095 | |||
DQ117585.1 | - | 2 | 2008 | |||
DQ117586.1 | - | 2 | 2008 | |||
DQ117587.1 | - | 2 | 2008 | |||
DQ117588.1 | - | 2 | 2008 | |||
DQ117589.1 | - | 2 | 2008 | |||
DQ117590.1 | - | 2 | 2008 | |||
DQ117594.1 | - | 2 | 2008 | |||
DQ117595.1 | - | 2 | 2008 | |||
DQ117596.1 | - | 2 | 2008 | |||
DQ117597.1 | - | 2 | 2008 | |||
DQ117598.1 | - | 2 | 2008 | |||
DQ117599.1 | - | 2 | 2008 | |||
DQ117600.1 | - | 2 | 2008 | |||
DQ153215.1 | - | 2 | 2008 | |||
DQ860103.1 | - | 2 | 2008 | |||
DQ860104.1 | - | 2 | 2008 |
