siRNA Details
Gene Symbol: Nlrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ117597.1 | - | 1 | 919 | ||
DQ117598.1 | - | 1 | 919 | |||
DQ117599.1 | - | 1 | 919 | |||
DQ153215.1 | - | 1 | 919 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nlrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ117597.1 | - | 1 | 919 | ||
DQ117598.1 | - | 1 | 919 | |||
DQ117599.1 | - | 1 | 919 | |||
DQ153215.1 | - | 1 | 919 |