siRNA Details
Gene Symbol: AW011738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036383.1 | - | 1 | 142 | ||
AK044300.1 | - | 143 | ||||
AK078660.1 | - | 1 | 143 | |||
AK083998.1 | - | 176 | ||||
AK085774.1 | - | 142 | ||||
AK086145.1 | - | 1 | 162 |
