siRNA Details
Gene Symbol: AW011738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036383.1 | - | 1 | 171 | ||
AK044300.1 | - | 172 | ||||
AK078660.1 | - | 1 | 172 | |||
AK083998.1 | - | 205 | ||||
AK085774.1 | - | 171 | ||||
AK086145.1 | - | 1 | 191 | |||
AK156747.1 | - | 1 | 175 | |||
AK172062.1 | - | 1 | 168 | |||
AK172315.1 | - | 1 | 168 |
