siRNA Details
Gene Symbol: AI467606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178901.3 | NP_849232.1 | 2 | 404 | ||
GenBank mRNA | AK041954.1 | - | 2 | 460 | ||
AK052739.1 | - | 2 | 405 | |||
AK155213.1 | - | 2 | 459 | |||
AK168260.1 | - | 2 | 406 | |||
AK169672.1 | - | 2 | 483 | |||
AK170238.1 | - | 2 | 416 | |||
BC038694.1 | - | 2 | 390 | 1513939 |
