siRNA Details
Gene Symbol: Ano10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271873.1 | NP_001258802.1 | 6 | 939 | ||
NM_133979.3 | NP_598740.1 | 6 | 1113 | |||
GenBank mRNA | AK028391.1 | - | 1 | 58 | ||
AK043011.1 | - | 6 | 1076 | |||
AK046091.1 | - | 6 | 1065 | |||
AK054163.1 | - | 2 | 403 | |||
AK079622.1 | - | 6 | 1064 | |||
AK080947.1 | - | 6 | 1065 | |||
AK138414.1 | - | 6 | 1188 | |||
AK188235.1 | - | 1 | 81 | |||
BC026421.1 | - | 1 | 153 | |||
BC099688.1 | - | 6 | 862 | 6313494 |
