siRNA Details
Gene Symbol: Slc6a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142809.1 | NP_001136281.1 | 8 | 1864 | ||
NM_001142810.1 | NP_001136282.1 | 8 | 1864 | |||
NM_133987.2 | NP_598748.1 | 8 | 1864 | |||
GenBank mRNA | AB077327.1 | - | 8 | 1212 | ||
AF459435.1 | - | 8 | 1847 | |||
AJ748650.1 | - | 5 | 454 | |||
AK189198.1 | - | 2 | 161 | |||
AK189504.1 | - | 5 | 284 | |||
BC024444.1 | - | 7 | 877 | |||
BC029000.1 | - | 6 | 627 | |||
BC049801.1 | - | 8 | 1039 | |||
BC141067.1 | - | 9 | 1605 | 9055679 | ||
BC145319.1 | - | 9 | 1269 | 9053874 | ||
EU523148.1 | - | 4 | 913 | |||
KC994641.1 | - | 4 | 913 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_017348, XM_006229611, BC168238.
Gene Ontology Categories:
