siRNA Details
Gene Symbol: Cep57l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243074.1 | NP_001230003.1 | 3 | 204 | ||
NM_001243075.1 | NP_001230004.1 | 2 | 123 | |||
NM_026643.5 | NP_080919.3 | 3 | 122 | |||
NM_029132.2 | NP_083408.1 | 3 | 190 | |||
GenBank mRNA | AF397908.1 | - | 3 | 118 | ||
AK010530.1 | - | 3 | 141 | |||
AK012710.1 | - | 2 | 69 | |||
AK015619.1 | - | 2 | 105 | |||
AK041460.1 | - | 2 | 91 | |||
BC138230.1 | - | 3 | 125 | 8861249 | ||
BQ553570.1 | - | 2 | 79 | |||
BY143469.1 | - | 2 | 91 |
