siRNA Details
Gene Symbol: Zfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034895.3 | NP_001030067.2 | 13 | 2020 | ||
GenBank mRNA | AK033649.1 | - | 6 | 769 | ||
AK122438.1 | - | 6 | 3628 | |||
BC072571.1 | - | 12 | 2689 | |||
BQ042879.1 | - | 1 | 41 |
