siRNA Details
Gene Symbol: Golm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035122.2 | NP_001030294.1 | 10 | 1229 | ||
NM_027307.4 | NP_081583.3 | 11 | 1365 | |||
GenBank mRNA | AK009072.1 | - | 9 | 1036 | ||
AK133865.1 | - | 10 | 1231 | |||
AK167646.1 | - | 10 | 1206 | |||
BC011152.1 | - | 10 | 1190 | 4188434 | ||
BC098486.1 | - | 11 | 1361 | 30024953 | ||
BU152451.1 | - | 2 | 107 | |||
CN529028.1 | - | 2 | 167 |
