siRNA Details
Gene Symbol: Dock9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081039.1 | NP_001074508.1 | 51 | 6044 | ||
NM_001128307.1 | NP_001121779.1 | 52 | 6347 | |||
NM_001128308.1 | NP_001121780.1 | 51 | 6308 | |||
NM_134074.2 | NP_598835.2 | 50 | 5894 | |||
GenBank mRNA | AK045750.1 | - | 21 | 2326 | ||
AK122431.1 | - | 52 | 6346 | |||
AK134685.1 | - | 4 | 552 | |||
BC009134.1 | - | 9 | 1232 | |||
BC046250.1 | - | 45 | 5351 | |||
BC052947.1 | - | 45 | 5363 | |||
BC057692.1 | - | 15 | 1876 | |||
BC157964.1 | - | 50 | 5914 | 9007289 | ||
BC158050.1 | - | 51 | 6239 | 9007354 | ||
BC158127.1 | - | 50 | 5777 |
