siRNA Details
Gene Symbol: Tecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027179.1 | NP_081455.1 | 4, 5 | 350 | ||
NM_134118.5 | NP_598879.1 | 5, 6 | 395 | |||
GenBank mRNA | AK002384.1 | - | 5, 6 | 365 | ||
AK010513.1 | - | 4, 5 | 286 | |||
AK010984.1 | - | 5, 6 | 369 | |||
AK013558.1 | - | 5, 6 | 351 | |||
AK020708.1 | - | 4, 5 | 344 | |||
BC019984.1 | - | 5, 6 | 381 | 4240045 | ||
BC094046.1 | - | 5, 6 | 495 | 5064002 | ||
BY278351.1 | - | 4, 5 | 350 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_138549, XM_006255218, XM_006255217, XM_006255216, XM_574311, and others.
Gene Ontology Categories:
