siRNA Details
Gene Symbol: Wdr90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163766.1 | NP_001157238.1 | 8 | 1229 | ||
GenBank mRNA | AK014328.1 | - | 4 | 330 | ||
AK036541.1 | - | 1 | 2258 | |||
AK129465.1 | - | 7 | 1144 | |||
AK154827.1 | - | 6 | 1396 | |||
AK170906.1 | - | 8 | 1189 |
