siRNA Details
Gene Symbol: Wdr90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163766.1 | NP_001157238.1 | 2 | 378 | ||
GenBank mRNA | AK036541.1 | - | 1 | 440 | ||
AK129465.1 | - | 1 | 347 | |||
AK170906.1 | - | 2 | 338 | |||
CJ102018.1 | - | 2 | 351 | |||
CN526398.1 | - | 2 | 90 |
