siRNA Details
Gene Symbol: Wdr90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163766.1 | NP_001157238.1 | 41 | 5643 | ||
GenBank mRNA | AK014328.1 | - | 37 | 4743 | ||
AK129465.1 | - | 40 | 5558 | |||
AK154827.1 | - | 39 | 5809 | |||
BB765139.1 | - | 2 | 86 | |||
BC009168.1 | - | 10 | 1295 | |||
BC025468.1 | - | 15 | 1901 | |||
BC043315.1 | - | 18 | 2334 |
