siRNA Details
Gene Symbol: Gpr137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177360.1 | NP_001170831.1 | 3 | 873 | ||
NM_001177361.1 | NP_001170832.1 | 5 | 1046 | |||
NM_001177362.1 | NP_001170833.1 | 5 | 737 | |||
NM_207220.2 | NP_997103.1 | 5 | 1149 | |||
GenBank mRNA | AK039612.1 | - | 5 | 649 | ||
AK142928.1 | - | 3 | 471 | |||
AK143450.1 | - | 5 | 1047 | |||
AK170308.1 | - | 5 | 739 | |||
BC048078.1 | - | 6 | 1021 | 6308873 |
