siRNA Details
Gene Symbol: Ece2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139293.2 | NP_647454.2 | 3 | 297 | ||
NM_177940.1 | NP_808809.1 | 3 | 517 | |||
NM_177941.1 | NP_808810.1 | 4 | 604 | |||
NM_177942.1 | NP_808811.1 | 2 | 210 | |||
GenBank mRNA | AF396699.1 | - | 3 | 238 | ||
AF489569.1 | - | 3 | 336 | |||
AF489570.1 | - | 4 | 423 | |||
AF489571.1 | - | 2 | 210 | |||
AF489572.1 | - | 3 | 297 | |||
AK031951.1 | - | 3 | 305 | |||
AU080537.1 | - | 3 | 254 | |||
BC138052.1 | - | 3 | 204 | 8861071 | ||
BC138053.1 | - | 3 | 204 | 8861072 | ||
BC145126.1 | - | 3 | 204 |
