siRNA Details
Gene Symbol: Rapgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039086.1 | NP_001034175.1 | 3, 4 | 668 | ||
NM_001039087.1 | NP_001034176.1 | 4, 5 | 782 | |||
NM_054050.2 | NP_473391.1 | 4, 5 | 782 | |||
GenBank mRNA | AF348669.1 | - | 4, 5 | 738 | ||
AK147468.1 | - | 4, 5 | 783 | |||
AK147470.1 | - | 4, 5 | 783 | |||
AK147506.1 | - | 4, 5 | 751 | |||
AK147696.1 | - | 3, 4 | 648 | |||
BC129881.1 | - | 3, 4 | 478 | 40110588 |
