siRNA Details
Gene Symbol: Ddb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028119.5 | NP_082395.2 | 8 | 1396 | ||
GenBank mRNA | AK011756.1 | - | 3 | 1187 | ||
AK158770.1 | - | 4 | 709 | |||
AY027937.1 | - | 8 | 1314 | |||
BC028955.1 | - | 7 | 1580 | |||
BC058589.1 | - | 5 | 1768 | |||
BC080821.1 | - | 7 | 1732 | |||
BC099410.1 | - | 4 | 596 | 30933658 | ||
BU515611.1 | - | 5 | 759 |
Gene Symbol: Acp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007387.2 | NP_031413.1 | 11 | 3043 | ||
GenBank mRNA | AK089401.1 | - | 11 | 3037 | ||
AK128963.1 | - | 11 | 3036 | |||
AK133756.1 | - | 11 | 2661 | |||
AK170658.1 | - | 11 | 3047 |

More Information
Other Species Targeted

Gene Ontology Categories:
