siRNA Details
Gene Symbol: Adamts6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081020.1 | NP_001074489.1 | 11 | 1792 | ||
GenBank mRNA | AK030315.1 | - | 12 | 2082 | ||
AK044528.1 | - | 12 | 1932 | |||
BC172116.1 | - | 12 | 1832 |
