siRNA Details
Gene Symbol: Mex3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175366.3 | NP_780575.2 | 2 | 1517 | ||
GenBank mRNA | AK077009.1 | - | 1 | 124 | ||
AK173330.1 | - | 1 | 2330 | |||
BC031512.1 | - | 1 | 501 | |||
BC067199.1 | - | 1 | 482 | |||
BC108401.1 | - | 1 | 725 |
