siRNA Details
Gene Symbol: Jmjd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242396.1 | NP_001229325.1 | 9 | 3308 | ||
NM_207221.2 | NP_997104.2 | 10 | 3076 | |||
GenBank mRNA | AK140633.1 | - | 10 | 3060 | ||
AK173162.1 | - | 9 | 3078 | |||
CD578529.1 | - | 3 | 422 |
