siRNA Details
Gene Symbol: Jmjd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242396.1 | NP_001229325.1 | 9 | 3779 | ||
NM_207221.2 | NP_997104.2 | 10 | 3547 | |||
GenBank mRNA | AK173162.1 | - | 9 | 3549 | ||
BC054559.1 | - | 1 | 110 | |||
BC068318.1 | - | 1 | 105 |
