siRNA Details
Gene Symbol: 1810012K08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007459.1 | - | 1 | 102 |
Gene Symbol: C330006A16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256521.1 | NP_001243450.1 | 2 | 632 | ||
NM_001256522.1 | NP_001243451.1 | 2 | 472 |
