siRNA Details
Gene Symbol: Tpbpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026429.4 | NP_080705.1 | 3 | 222 | ||
GenBank mRNA | AK005433.1 | - | 3 | 222 | ||
AK018793.1 | - | 3 | 221 | |||
AK018794.1 | - | 3 | 220 | |||
AK018795.1 | - | 3 | 222 | |||
AK018798.1 | - | 3 | 222 | |||
AK018799.1 | - | 3 | 222 | |||
AK018804.1 | - | 3 | 223 | |||
AK018805.1 | - | 3 | 222 | |||
AK018808.1 | - | 3 | 223 | |||
AK018809.1 | - | 3 | 221 | |||
AK018810.1 | - | 3 | 222 | |||
AK018817.1 | - | 3 | 222 | |||
AK018820.1 | - | 3 | 222 | |||
AK018824.1 | - | 3 | 222 | |||
AK018827.1 | - | 3 | 221 | |||
AY034576.1 | - | 3 | 197 | |||
BC099441.1 | - | 3 | 207 | |||
BC100333.1 | - | 3 | 207 | 30787126 |
