siRNA Details
Gene Symbol: 1110038F14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054099.2 | NP_473440.1 | 4 | 730 | ||
GenBank mRNA | AK004154.1 | - | 4 | 684 | ||
AK141496.1 | - | 4 | 522 | |||
AK153059.1 | - | 5 | 684 | |||
AK160695.1 | - | 4 | 753 | |||
AK185822.1 | - | 1 | 23 | |||
BC038324.1 | - | 4 | 730 | 1511597 | ||
BC091734.1 | - | 4 | 495 | 3663572 |
