siRNA Details
Gene Symbol: Ap2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077264.1 | NP_001070732.1 | 13, 14 | 1924 | ||
NM_007458.2 | NP_031484.1 | 13, 14 | 1924 | |||
GenBank mRNA | AK135424.1 | - | 13, 14 | 1853 | ||
AK155515.1 | - | 13, 14 | 1861 | |||
BC031433.1 | - | 13, 14 | 1874 | 4481643 |
