siRNA Details
Gene Symbol: Cirbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007705.2 | NP_031731.1 | 4 | 323 | ||
GenBank mRNA | AK014655.1 | - | 4 | 326 | ||
AK132584.1 | - | 4 | 326 | |||
BC075699.1 | - | 4 | 313 | 30656047 | ||
D78135.1 | - | 4 | 323 |
