siRNA Details
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007742.3 | NP_031768.2 | 48 | 3780 | ||
GenBank mRNA | AK132180.1 | - | 48 | 3802 | ||
AK159283.1 | - | 48 | 3797 | |||
AK159285.1 | - | 48 | 3802 | |||
BC003198.1 | - | 11 | 1090 | |||
BC042642.1 | - | 41 | 3350 | |||
BC050014.1 | - | 48 | 3780 | 4225038 | ||
BC059281.1 | - | 41 | 3106 | 3992363 | ||
U03419.1 | - | 2 | 297 | |||
U08020.1 | - | 48 | 3681 |
