siRNA Details
Gene Symbol: Dab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008702.2 | NP_001008702.1 | 5, 6 | 682 | ||
NM_001037905.3 | NP_001032994.1 | 5, 6 | 756 | |||
NM_001102400.1 | NP_001095870.1 | 5, 6 | 756 | |||
NM_023118.5 | NP_075607.2 | 5, 6 | 756 | |||
GenBank mRNA | AK002850.1 | - | 5, 6 | 639 | ||
AK154716.1 | - | 5, 6 | 740 | |||
AK162744.1 | - | 5, 6 | 727 | |||
AK162851.1 | - | 5, 6 | 727 | |||
AK165519.1 | - | 5, 6 | 640 | |||
AK165810.1 | - | 5, 6 | 637 | |||
AK168375.1 | - | 6 | 730 | |||
BC006588.1 | - | 4, 5 | 577 | 3587226 | ||
BC016887.1 | - | 5, 6 | 622 | 4242174 | ||
CD348957.1 | - | 3, 4 | 279 |
