siRNA Details
Gene Symbol: Cd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010016.2 | NP_034146.2 | 7 | 951 | ||
GenBank mRNA | AK030285.1 | - | 7 | 943 | ||
AK160994.1 | - | 7 | 929 | |||
BC011314.1 | - | 7 | 906 | 3663561 | ||
CT010328.1 | - | 7 | 880 | |||
D63679.1 | - | 7 | 862 | |||
L41366.1 | - | 7 | 895 |
