siRNA Details
Gene Symbol: Dapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010019.3 | NP_034149.2 | 8, 9 | 934 | ||
GenBank mRNA | AB018002.1 | - | 8, 9 | 924 | ||
BC022165.1 | - | 8, 9 | 903 | 4016911 | ||
BX527735.1 | - | 4, 5 | 318 | |||
GU109477.1 | - | 8, 9 | 934 |
