siRNA Details
Gene Symbol: Dclk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111051.1 | NP_001104521.1 | 2 | 346 | ||
NM_001111052.1 | NP_001104522.1 | 2 | 346 | |||
NM_001111053.1 | NP_001104523.1 | 6 | 1207 | |||
NM_001195538.1 | NP_001182467.1 | 6 | 1207 | |||
NM_001195539.1 | NP_001182468.1 | 2 | 346 | |||
NM_001195540.1 | NP_001182469.1 | 2 | 346 | |||
NM_019978.3 | NP_064362.1 | 6 | 1207 | |||
GenBank mRNA | AB073969.1 | - | 2 | 154 | ||
AB093234.1 | - | 6 | 1164 | |||
AF155819.1 | - | 6 | 1105 | |||
AF155820.1 | - | 6 | 1146 | |||
AF155821.1 | - | 2 | 205 | |||
AK032424.1 | - | 2 | 188 | |||
AK043874.1 | - | 6 | 1128 | |||
AK047716.1 | - | 2 | 133 | |||
AK081200.1 | - | 2 | 189 | |||
AK147831.1 | - | 6 | 1207 | |||
AK164993.1 | - | 6 | 1165 | |||
AY968047.1 | - | 2 | 97 | |||
AY968048.1 | - | 2 | 97 | |||
BC021354.1 | - | 6 | 1149 | |||
BC050903.1 | - | 6 | 1098 | |||
BC064783.1 | - | 6 | 1154 | |||
BC133685.1 | - | 6 | 1009 | 40110683 | ||
BM933369.1 | - | 2 | 339 |
