siRNA Details
Gene Symbol: Dpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159543.1 | NP_001153015.1 | 7 | 861 | ||
NM_010074.3 | NP_034204.1 | 7 | 861 | |||
GenBank mRNA | AK040409.1 | - | 6 | 567 | ||
AK085370.1 | - | 7 | 444 | |||
AK153939.1 | - | 7 | 475 | |||
AK163085.1 | - | 7 | 866 | |||
AK164775.1 | - | 7 | 444 | |||
BC022183.1 | - | 7 | 723 | 3982742 | ||
X58384.1 | - | 7 | 538 |
