siRNA Details
Gene Symbol: Plxnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019587.2 | NP_062533.2 | 24 | 4233 | ||
GenBank mRNA | AK048728.1 | - | 8 | 1715 | ||
AK122467.1 | - | 19 | 3565 | |||
AK138168.1 | - | 20 | 3238 | |||
AK138631.1 | - | 19 | 3518 | |||
AK140240.1 | - | 22 | 4415 | |||
AK170004.1 | - | 11 | 1483 |
