siRNA Details
Gene Symbol: Srgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081011.2 | NP_001074480.2 | 3 | 964 | ||
GenBank mRNA | AK084412.1 | - | 2 | 843 | ||
AK143649.1 | - | 3 | 955 | |||
BC030457.1 | - | 2 | 271 | |||
BC151081.1 | - | 2 | 582 | 9087994 | ||
BC151082.1 | - | 2 | 582 | 9087995 | ||
BC158055.1 | - | 2 | 580 | 9088163 | ||
BC172152.1 | - | 2 | 582 | 9054822 | ||
CF164437.1 | - | 2 | 241 |
Gene Symbol: Cdk5rap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC027761.1 | - | 3402 |
