siRNA Details
Gene Symbol: Fxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008044.2 | NP_032070.1 | 3 | 353 | ||
GenBank mRNA | AK148147.1 | - | 3 | 375 | ||
AK160458.1 | - | 3 | 363 | |||
BC016644.1 | - | 2 | 1066 | |||
BC038010.1 | - | 2 | 1077 | |||
BC058533.1 | - | 3 | 335 | |||
BC125337.1 | - | 3 | 351 | 40129852 | ||
BC132337.1 | - | 3 | 351 | 40130614 | ||
U95736.1 | - | 3 | 353 |
