siRNA Details
Gene Symbol: Galr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015738.2 | NP_056553.2 | 1 | 329 | ||
GenBank mRNA | BC131950.1 | - | 1 | 329 | 40130227 | |
BC131952.1 | - | 1 | 329 | 40130229 |
Gene Symbol: Gcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161712.1 | NP_001155184.1 | 9 | 1445 | ||
GenBank mRNA | AK076469.1 | - | 9 | 1418 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
