siRNA Details
Gene Symbol: Galr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015738.2 | NP_056553.2 | 2 | 905 | ||
GenBank mRNA | BC131950.1 | - | 2 | 905 | 40130227 | |
BC131952.1 | - | 2 | 905 | 40130229 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
