siRNA Details
Gene Symbol: Glul | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008131.4 | NP_032157.2 | 6, 7 | 939 | ||
GenBank mRNA | AK137969.1 | - | 6, 7 | 926 | ||
AK159106.1 | - | 6, 7 | 925 | |||
AK160670.1 | - | 6, 7 | 926 | |||
AK162685.1 | - | 6, 7 | 924 | |||
AK168493.1 | - | 6, 7 | 923 | |||
AY044241.1 | - | 6, 7 | 923 | |||
BC015086.1 | - | 6, 7 | 923 | 4224865 | ||
X16314.1 | - | 6, 7 | 900 |
