siRNA Details
Gene Symbol: Gnat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008141.3 | NP_032167.1 | 8 | 872 | ||
GenBank mRNA | AK009388.1 | - | 7 | 756 | ||
AK028395.1 | - | 4 | 389 | |||
AY974584.1 | - | 1 | 25 | |||
BC016272.1 | - | 8 | 870 | 4506728 | ||
BC085102.1 | - | 6 | 1619 | |||
BC131941.1 | - | 8 | 867 | 40130218 | ||
BC131943.1 | - | 8 | 867 | 40130220 | ||
L10666.1 | - | 7 | 794 |
