siRNA Details
Gene Symbol: Gnaz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010311.3 | NP_034441.1 | 4 | 1611 | ||
GenBank mRNA | AF056972.1 | - | 2 | 546 | ||
AK080716.1 | - | 4 | 1611 | |||
AK133483.1 | - | 2 | 1487 | |||
AK134710.1 | - | 2 | 1315 | |||
BC014702.1 | - | 2 | 1312 | 4501587 |
