siRNA Details
Gene Symbol: Hba-x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010405.4 | NP_034535.1 | 2 | 296 | ||
GenBank mRNA | AK077590.1 | - | 2 | 295 | ||
BC051988.1 | - | 2 | 266 | 5711898 | ||
BY111754.1 | - | 2 | 297 | |||
M13125.1 | - | 1 | 104 | |||
M26898.1 | - | 2 | 254 |
