siRNA Details
Gene Symbol: Hfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010424.4 | NP_034554.2 | 3 | 539 | ||
GenBank mRNA | AK009581.1 | - | 3 | 540 | ||
AK088986.1 | - | 3 | 535 | |||
AK150697.1 | - | 3 | 557 | |||
BC116744.1 | - | 3 | 497 | 40126063 | ||
BC116746.1 | - | 3 | 495 | 40126065 | ||
U66849.1 | - | 3 | 521 |
