siRNA Details
Gene Symbol: Hoxa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008265.3 | NP_032291.1 | 2 | 859 | ||
GenBank mRNA | AK076098.1 | - | 2 | 860 | ||
BC107172.2 | - | 3 | 566 | 40044111 | ||
M27432.1 | - | 1 | 307 | |||
X13538.1 | - | 1 | 306 |
