siRNA Details
Gene Symbol: Ids | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010498.3 | NP_034628.2 | 9 | 1648 | ||
GenBank mRNA | AJ627044.1 | - | 7 | 1113 | ||
AK085237.1 | - | 10 | 1525 | |||
AK132287.1 | - | 10 | 1528 | |||
AK166178.1 | - | 9 | 1526 | |||
AK166443.1 | - | 10 | 1477 |
