siRNA Details
Gene Symbol: Irak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177973.1 | NP_001171444.1 | 7 | 1027 | ||
NM_001177974.1 | NP_001171445.1 | 7 | 1024 | |||
NM_001177975.1 | NP_001171446.1 | 7 | 1024 | |||
NM_001177976.1 | NP_001171447.1 | 7 | 1024 | |||
NM_008363.2 | NP_032389.2 | 7 | 1024 | |||
GenBank mRNA | AB088370.1 | - | 7 | 851 | ||
AF103876.1 | - | 7 | 1024 | |||
AK045958.1 | - | 7 | 933 | |||
AK143694.1 | - | 7 | 935 | |||
AK144879.1 | - | 7 | 933 | |||
AK210148.1 | - | 3 | 291 | |||
AY184362.1 | - | 7 | 1024 | |||
AY184363.1 | - | 7 | 1024 | |||
AY184364.1 | - | 7 | 1024 | |||
BC004778.1 | - | 7 | 1019 | 3257579 | ||
U56773.2 | - | 7 | 848 |
