siRNA Details
Gene Symbol: Fabp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008375.2 | NP_032401.1 | 1, 2 | 128 | ||
GenBank mRNA | BC119289.1 | - | 1, 2 | 98 | 8734038 | |
BC120767.1 | - | 1, 2 | 98 | 40129690 | ||
CJ043022.1 | - | 1, 2 | 128 |
