siRNA Details
Gene Symbol: Gimap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008376.3 | NP_032402.2 | 3 | 659 | ||
NM_175860.2 | NP_787056.1 | 3 | 653 | |||
GenBank mRNA | AK156732.1 | - | 3 | 654 | ||
AK169133.1 | - | 3 | 612 | |||
BC132601.1 | - | 3 | 619 | 40130878 | ||
BC132603.1 | - | 3 | 619 | 40130880 | ||
BY751570.1 | - | 3 | 654 | |||
Y08026.2 | - | 1 | 768 |
