siRNA Details
Gene Symbol: Irx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008393.3 | NP_032419.2 | 2, 3 | 1804 | ||
GenBank mRNA | BC067195.1 | - | 2, 3 | 1803 | ||
BC085500.1 | - | 2, 3 | 1524 | 30539191 | ||
Y15001.1 | - | 2, 3 | 1390 |
